missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MATR3 Recombinant Rabbit Monoclonal Antibody (24GB535)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557465
This item is not returnable.
View return policy
Description
MATR3 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene encodes a nuclear matrix protein, which is proposed to stabilize certain messenger RNA species. Mutations of this gene are associated with distal myopathy 2, which often includes vocal cord and pharyngeal weakness. Alternatively spliced transcript variants, including read-through transcripts composed of the upstream small nucleolar RNA host gene 4 (non-protein coding) and matrin 3 gene sequence, have been identified. Pseudogenes of this gene are located on chromosomes 1 and X.
Specifications
| MATR3 | |
| Recombinant Monoclonal | |
| 1.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P43243, P43244, Q8K310 | |
| MATR3 | |
| A synthesized peptide derived from human Matrin 3 (aa 750-800). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB535 | |
| Unconjugated | |
| MATR3 | |
| 1110061A14Rik; 2810017I02Rik; AI841759; ALS21; AW555618; D030046F20Rik; KIAA0723; MATR3; matrin 3; matrin-3; matrin-3-like protein; mKIAA0723; MPD2; nuclear scaffold protein P130/MAT3; P130/MAT3; VCPDM; vocal cord and pharyngeal weakness with distal myopathy | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 17184, 29150, 9782 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction