missing translation for 'onlineSavingsMsg'
Learn More

MATN3 Antibody (391934) [mFluor Violet 450 SE], Novus Biologicals™

Product Code. 30731484 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30731484 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30731484 Supplier Novus Biologicals Supplier No. FAB3017MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MATN3 Monoclonal antibody specifically detects MATN3 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen MATN3
Applications Western Blot
Classification Monoclonal
Clone 391934
Conjugate mFluor Violet 450 SE
Formulation 50mM Sodium Borate
Gene Alias DIPOA, EDM5, HOA, matrilin 3, matrilin-3, OADIP, OS2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Matrilin-3, Asp29-Arg486, Accession # O15232
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4148
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.