missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MATN2 Polyclonal Antibody

Description
A suggested positive control is human liver tissue lysate. PA5-20734 can be used with blocking peptide PEP-0848.
Matrilin (MATNs) are a family of non-collagenous extracellular matrix proteins consisting of four known members that have been proposed to play key roles in the formation of both collagen-dependent and collagen-independent filamentous networks. The matrilin family all share a structure made up of von Willebrand factor A domains, epidermal growth factor-like domains and a coiled coil alpha-helical module. MATN1 and MATN3 are expressed mainly in cartilage, while MATN2 and MATN4 occur in a wide variety of extracellular matrices. The matrilin genes are strictly and differently regulated and their expression may serve as markers for cellular differentiation and diseases such as astrocytoma and liver carcinoma. Recent studies show that MATN2 is a permissive substrate for axonal growth and cell migration, and it is required for successful nerve regeneration.
Specifications
Specifications
| Antigen | MATN2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | MATN2 |
| Gene Accession No. | O00339, O08746 |
| Gene Alias | Crtm2; MATN2; matrilin 2; matrilin-2; testis tissue sperm-binding protein Li 94mP; UNQ193/PRO219 |
| Gene Symbols | MATN2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?