missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MATN1 Polyclonal Antibody

Product Code. 12808872
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12808872 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12808872 Supplier Invitrogen™ Supplier No. PA520725

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is mouse liver tissue lysate. PA5-20725 can be used with blocking peptide PEP-0839.

This gene catalyzes a two-step reaction that involves the transfer of the adenosyl moiety of ATP to methionine to form S-adenosylmethionine and tripolyphosphate, which is subsequently cleaved to PPi and Pi. S-adenosylmethionine is the source of methyl groups for most biological methylations. The encoded protein is found as a homotetramer or a homodimer whereas a third form, MAT II , is encoded by the MAT2A gene. Mutations in this gene are associated with methionine adenosyltransferase deficiency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MATN1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene MATN1
Gene Accession No. P21941, P51942
Gene Alias cartilage matrix protein; Cmp; CRTM; MAT; Mat1; MAT1A; MATA1; MATN1; matrilin 1, cartilage matrix protein; matrilin 1, cartilage matrix protein 1; matrilin-1; SAMS; SAMS1
Gene Symbols MATN1
Host Species Rabbit
Immunogen a 12 amino acid peptide from near the amino terminus of human MATN1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17180, 297894, 4146
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.