missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MATK Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA577272
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
MATK (Hyl) is a non-receptor CSK-type protein tyrosine kinase that is thought to function in signal transduction pathways important in megakaryocyte growth and/or differentiation. This protein is thought to play a significant role in the signal transduction of hematopoietic cells. It is able to phosphorylate and inactivate Src family kinases, and may play an inhibitory role in the control of T-cell proliferation. This protein might be involved in signaling in some cases of breast cancer.
Specifications
| MATK | |
| Polyclonal | |
| Unconjugated | |
| MATK | |
| Batk; CHK; Csk homologous kinase; Csk-homologous kinase; Csk-type protein tyrosine kinase; CTK; DKFZp434N1212; Hematopoietic consensus tyrosine-lacking kinase; HHYLTK; hydroxyaryl-protein kinase; HYL; HYL tyrosine kinase; HYLTK; leukocyte carboxyl-terminal src kinase related; Lsk; Matk; megakaryocyte-associated tyrosine kinase; megakaryocyte-associated tyrosine kinase (non-receptor protein tyrosine kinase); megakaryocyte-associated tyrosine-protein kinase; MGC1708; MGC2101; non-receptor protein kinase protein; Ntk; Protein kinase BATK; protein kinase HYL; Protein kinase NTK; tyrosine kinase MATK; tyrosine-protein kinase CTK; tyrosylprotein kinase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 17179, 4145, 60450 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide; pH 7.2 | |
| P41242, P41243, P42679 | |
| MATK | |
| Synthetic peptide corresponding to amino acids 140-180 of Human MATK. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction