missing translation for 'onlineSavingsMsg'
Learn More

MATH2/NEUROD6 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-29865

 View more versions of this product

Product Code. 18142494

  • £649.00 / 0.10mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MATH2/NEUROD6 Polyclonal specifically detects MATH2/NEUROD6 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MATH2/NEUROD6
Polyclonal
Western Blot 1:200-1:1000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin
Q96NK8
NEUROD6
Synthetic peptide.
0.1 mg
Primary
Recognizes NeuroD 6 (MATH2, NEX1), a Helix-loop-helix class of transcription factor.
Store at -70°C. Avoid freeze-thaw cycles.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
50% saturated ammonium sulfate and PBS with No Preservative
Atoh2, BHLHA2, bHLHa2MATH2, Class A basic helix-loop-helix protein 2, Math-2, NeuroD6, neurogenic differentiation 6, neurogenic differentiation factor 6, NEX1M, Protein atonal homolog 2
Rabbit
Affinity Purified
RUO
63974
Human, Mouse
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only