missing translation for 'onlineSavingsMsg'
Learn More

MATH2/NEUROD6 Antibody (3B3), Novus Biologicals™

Product Code. 18419138 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18419138 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18419138 Supplier Novus Biologicals Supplier No. H00063974M06

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MATH2/NEUROD6 Monoclonal antibody specifically detects MATH2/NEUROD6 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen MATH2/NEUROD6
Applications Western Blot, ELISA, Immunofluorescence
Classification Monoclonal
Clone 3B3
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_073565
Gene Alias Atoh2, BHLHA2, bHLHa2MATH2, Class A basic helix-loop-helix protein 2, Math-2, NeuroD6, neurogenic differentiation 6, neurogenic differentiation factor 6, NEX1M, Protein atonal homolog 2
Host Species Mouse
Immunogen NEUROD6 (NP_073565.2, 191 a.a. ∽ 290 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. GQGGEAAHHTRSPYSTFYPPYHSPELTTPPGHGTLDNSKSMKPYNYCSAYESFYESTSPECASPQFEGPLSPPPINYNGIFSLKQEETLDYGKNYNYGMH
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63974
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.