missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MATH1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-31318
This item is not returnable.
View return policy
Description
MATH1 Polyclonal antibody specifically detects MATH1 in Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.
Specifications
| MATH1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 20% Glycerol (pH7) with 0.025% Proclin 300 | |
| ATH1protein atonal homolog 1, atonal homolog 1 (Drosophila), BHLHA14, bHLHa14Math1, Class A basic helix-loop-helix protein 14, hATH1, Helix-loop-helix protein hATH-1, MATH-1 | |
| Rabbit | |
| 38 kDa | |
| 0.1 mL | |
| Cancer | |
| 474 | |
| Mouse, Rat | |
| IgG |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.75mg/mL | |
| Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
| Q92858 | |
| ATOH1 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human MATH1. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse. Predicted cross-reactivity based on sequence identity: Chimpanzee (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction