missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAT2B Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558074
This item is not returnable.
View return policy
Description
Immunogen sequence: AVGAFLIYIS SDYVFDGTNP PYREEDIPAP LNLYGKTKLD GEKAVLENNL GAAVLRIPIL YGEVEKLEES AVTVM Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
MAT2B belongs to the methionine adenosyltransferase (MAT) family. MAT catalyzes the biosynthesis of S-adenosylmethionine from methionine and ATP. This protein is the regulatory beta subunit of MAT. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified.
Specifications
| MAT2B | |
| Polyclonal | |
| Unconjugated | |
| MAT2B | |
| 1110064C04Rik; 2410018D16Rik; AI182287; AU022853; beta regulatory subunit of methionine adenosyltransferase; dTDP-4-keto-6-deoxy-D-glucose 4-reductase; MAT II beta; MAT2B; MAT-II; MATIIbeta; methionine adenosyltransferase 2 beta subunit; methionine adenosyltransferase 2 subunit beta; methionine adenosyltransferase 2B; Methionine adenosyltransferase II beta; methionine adenosyltransferase II, beta; MSTP045; Nbla02999; Putative dTDP-4-keto-6-deoxy-D-glucose 4-reductase; putative protein product of Nbla02999; SDR23E1; short chain dehydrogenase/reductase family 23E, member 1; testicular tissue protein Li 118; TGR; UNQ2435/PRO4995 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 27430 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NZL9 | |
| MAT2B | |
| Recombinant protein corresponding to Human MAT2B. Recombinant protein control fragment (Product #RP-96792). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction