Learn More
Invitrogen™ MAT2B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530530
Description
Recommended positive controls: 293T. Predicted reactivity: Human (99%), Mouse (96%), Rat (97%), Xenopus laevis (80%), Pig (98%), Rhesus Monkey (99%), Chimpanzee (99%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
MAT2B belongs to the methionine adenosyltransferase (MAT) family. MAT catalyzes the biosynthesis of S-adenosylmethionine from methionine and ATP. This protein is the regulatory beta subunit of MAT. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified.
Specifications
| MAT2B | |
| Polyclonal | |
| Unconjugated | |
| MAT2B | |
| 1110064C04Rik; 2410018D16Rik; AI182287; AU022853; beta regulatory subunit of methionine adenosyltransferase; dTDP-4-keto-6-deoxy-D-glucose 4-reductase; MAT II beta; MAT2B; MAT-II; MATIIbeta; methionine adenosyltransferase 2 beta subunit; methionine adenosyltransferase 2 subunit beta; methionine adenosyltransferase 2B; Methionine adenosyltransferase II beta; methionine adenosyltransferase II, beta; MSTP045; Nbla02999; Putative dTDP-4-keto-6-deoxy-D-glucose 4-reductase; putative protein product of Nbla02999; SDR23E1; short chain dehydrogenase/reductase family 23E, member 1; testicular tissue protein Li 118; TGR; UNQ2435/PRO4995 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 108645, 27430 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.45 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q99LB6, Q9NZL9 | |
| MAT2B | |
| Recombinant protein encompassing a sequence within the center region of human MAT2B. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.