missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAT2B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35174-100ul
This item is not returnable.
View return policy
Description
MAT2B Polyclonal antibody specifically detects MAT2B in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| MAT2B | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| beta regulatory subunit of methionine adenosyltransferase, DTDP-4-keto-6-deoxy-D-glucose 4-reductase, MAT II beta, MAT-II, MATIIbeta, methionine adenosyltransferase 2 subunit beta, Methionine adenosyltransferase II beta, methionine adenosyltransferase II, beta, MGC12237, Nbla02999, putative protein product of Nbla02999, SDR23E1, short chain dehydrogenase/reductase family 23E, member 1, TGR | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-334 of human MAT2B (NP_037415.1).,, Sequence:, MVGREKELSIHFVPGSCRLVEEEVNIPNRRVLVTGATGLLGRAVHKEFQQNNWHAVGCGFRRARPKFEQVNLLDSNAVHHIIHDFQPHVIVHCAAERRPDVVENQPDAASQLNVDASGNLAKEAAAVGAFLIYISSDYVFDGTNPPYREEDIPAPLNLYGKTKLDGEKAVLENNLGAAVLRIPILYGEVEKLEESAVTVMFDKVQFSNKSANMDHWQQRFPTHVKDVATVCRQLAEKRMLDPSIKGTFHWSGNEQMTKYEMACAIADAFNLPSSHLRPITDSPVLGAQRPRNAQLDCSKLETLGIGQRTPFRIGIKESLWPFLIDKRWRQTVFH | |
| 100 μL | |
| Stem Cell Markers | |
| 27430 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction