Learn More
MAT1/2A Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-94162PE
This item is not returnable.
View return policy
Description
MAT1/2A Polyclonal antibody specifically detects MAT1/2A in Human, Rat, Bovine, Primate, Zebrafish, Mouse (Negative) samples. It is validated for Western Blot, Western Blot
Specifications
| Western Blot |