missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MASTL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584190
This item is not returnable.
View return policy
Description
Immunogen sequence: EVLKTLASKR NAVAFRSFNS HINASNNSEP SRMNMTSLDA MDISCAYSGS YPMAITPTQK RRSCMPHQTP NQIKSG Highest antigen sequence indentity to the following orthologs: Mouse - 70%, Rat - 68%.
MASTL or microtubule associated serine/threonine kinase-like belongs to the MAST family, AGC Ser/Thr protein kinase group. MASTL promotes mitotic progression and cell cycle reentry after DNA damage. Loss of MASTL leads to defects in chromosome condensation, separation, and other aspects of mitotic progression, which is primarily mediated by inhibiting PP2A/B55 delta. Overexpression of MASTL is correlated with cancer progression, poor patient survival, and tumor recurrence in various cancers. Mutations at this locus have been associated with autosomal dominant thrombocytopenia, also known as thrombocytopenia-2. Alternatively spliced transcript variants have been described for this locus.
Specifications
| MASTL | |
| Polyclonal | |
| Unconjugated | |
| MASTL | |
| 2700091H24Rik; C88295; GREATWALL; greatwall kinase homolog; greatwall protein kinase; GW; GWL; hGWL; Mastl; MAST-L; microtubule associated serine/threonine kinase like; microtubule associated serine/threonine kinase-like; microtubule-associated serine/threonine-protein kinase-like; RP11-85G18.2; Serine/threonine-protein kinase greatwall; THC2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84930 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96GX5 | |
| MASTL | |
| Recombinant protein corresponding to Human MASTL. Recombinant protein control fragment (Product #RP-106519). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction