missing translation for 'onlineSavingsMsg'
Learn More

MASTL Antibody, DyLight 350, Novus Biologicals™

Product Code. 18420554 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18420554 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18420554 Supplier Novus Biologicals Supplier No. NBP122987UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MASTL Polyclonal antibody specifically detects MASTL in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen MASTL
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 350
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias FLJ14813EC 2.7.11.1, GREATWALL, GW, GWL, hGWL, MAST-L, microtubule associated serine/threonine kinase-like, Microtubule-associated serine/threonine-protein kinase-like, RP11-85G18.2, THC2greatwall protein kinase
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 550 and 600 of human microtubule associated serine/threonine kinase-like using the numbering given in entry NP_116233.2 (GeneID 84930).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 84930
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.