Learn More
BarbLock Assemblies
Product Code. 15272700
Click to view available options
For Use With:
Fits 1/8 in. Tubing
Fits 1/8 in. x 1/4 in. Tubing
Unit Size:
Each
Product Code. 15272700
Brand: Masterflex 3060418
Only null left
Add to basket
Add to basket
Specifications
For Use With Fits 1/8 in. Tubing
For Use With (Application) Easy to install, three-piece locking mechanism ensures the tubing and hose barb won't disengage
Packaging Quantity 1 Pack
Product Type BarbLock Assembly