Learn More
Eppendorf™ Mastercycler X50P, GB-Plug
The elegant synthesis of speed, flexibility and PCR optimization for research PCR applications and standardization for routine PCR applications.
Brand:  Eppendorf™ 6315000040
Product Code. 15778419
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Heating rate: up to 10°C/s
  • Wide selection of blocks from a fast silver block to 384 well block
  • Touch screen interface
  • Innovative 2D-Gradient for advanced PCR optimization
  • X50 with Touchscreen can control up to 9 other units (connected cyclers do not have a touch screen interface)
  • flexlid™ concept: automatic height adjustment of the lid allows you to use all types of consumables
  • Small footprint
  • 2-year warranty