missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One Masterblock™ 96-well Microplates
GREENER_CHOICE

Product Code. 10090292
Change view
Click to view available options
Total Volume (Metric) Well:
0.5 mL
Unit Size:
Pack of 80
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Total Volume (Metric) Well Unit Size
10090292 0.5 mL Pack of 80
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10090292 Supplier Greiner Bio-One Supplier No. 786201
Estimated Shipment: 17-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Manufactured from polypropylene for excellent heat stability and chemical resistance. Greiner Bio One™ Masterblock™ 96 Well 0.5mL Microplates provide a unique deep well design suitable for applications where larger working volumes are required. The plate is compatible with current plate equipment and can be used for manual and robotic handling.

  • Footprint compatible with automated systems
  • Can be traced to production through defined LOT number system
  • ideal for storage of active agents, patient samples, biomolecules and reagents and cultivation of bacteria or yeasts
  • Uniform external dimensions and tolerances
  • Alphnumeric well coding
  • Free of detectable DNase, RNase and human DNA
  • Non-pyrogenic
  • Manufactured under DIN ISO 9001 guidelines
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance DIN ISO 9001
Certified RNase-/DNase, pyrogen free
Format Standard
Array 8 x 12
Color Natural
Material Polypropylene
No. of Wells 96
Sterility Non-sterile
Total Volume (Metric) Well 0.5 mL
Well Shape Conical Bottom (V-well)
Dnase Free Yes
No. per Pack 80/Cs
Profile V-bottom
Quantity 80/Cs.
RNase Free Yes
Temperature Range (Metric) -196°C to +121°C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.