missing translation for 'onlineSavingsMsg'
Learn More

Atago™ MASTER-SUR/Nα Model Clinical Refractometer

Product Code. 11466168
Change view
Click to view available options
Refractive Index:
1.333 to 1.356
1.3330 to 1.3660
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Refractive Index unitSize
11466168 1.333 to 1.356 Each
11446168 1.3330 to 1.3660 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11466168 Supplier Atago™ Supplier No. 2791

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Type Clinical Refractometer
Scale Range 1.000 to 1.060 Urine Specific Gravity, 0 to 12 g/100 mL Serum Protein
Refractive Index 1.333 to 1.356
Model MASTER-URC/Nα
Accuracy ±0.001 Urine S.G. Scale, ±0.0005nD at (10 to 30°C) Refractive Index Scale
Scale Type Urine S.G., Refractive Index
For Use With (Application) Designed to quickly and easily measure Urine Specific Gravity with just a single drop of sample
IP Rating IP65
Weight (Metric) 110 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.