missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAST205 Antibody (OTI3A5), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72592AF750
This item is not returnable.
View return policy
Description
MAST205 Monoclonal specifically detects MAST205 in Human, Mouse, Rat samples. It is validated for Western Blot, Knockout Validated.
Specifications
| MAST205 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| MAST2 | |
| Recombinant protein fragment corresponding to amino acids 1482-1798 of human MAST205 (NP_055927) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunoassay | |
| OTI3A5 | |
| Western Blot, Knockout Validated | |
| EC 2.7.11, EC 2.7.11.1, FLJ39200, KIAA0807RP4-533D7.1, MAST205microtubule associated testis specific serine/threonine protein kinase, microtubule associated serine/threonine kinase 2, microtubule-associated serine/threonine-protein kinase 2, MTSSK | |
| Mouse | |
| Affinity Purified | |
| Protein Kinase | |
| 23139.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction