missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MAST205 Polyclonal antibody specifically detects MAST205 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | MAST205 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 680 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EC 2.7.11, EC 2.7.11.1, FLJ39200, KIAA0807RP4-533D7.1, MAST205microtubule associated testis specific serine/threonine protein kinase, microtubule associated serine/threonine kinase 2, microtubule-associated serine/threonine-protein kinase 2, MTSSK |
| Host Species | Rabbit |
| Immunogen | The immunogen recognized by this antibody maps to a region between residue 25 and 75 of human microtubule associated serine/threonine kinase 2 using the numbering given in entry NP_055927.2 (GeneID 23139). |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?