missing translation for 'onlineSavingsMsg'
Learn More

MAST205 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18407262 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407262 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407262 Supplier Novus Biologicals Supplier No. NBP122985AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MAST205 Polyclonal antibody specifically detects MAST205 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAST205
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.11, EC 2.7.11.1, FLJ39200, KIAA0807RP4-533D7.1, MAST205microtubule associated testis specific serine/threonine protein kinase, microtubule associated serine/threonine kinase 2, microtubule-associated serine/threonine-protein kinase 2, MTSSK
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 25 and 75 of human microtubule associated serine/threonine kinase 2 using the numbering given in entry NP_055927.2 (GeneID 23139).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 23139
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.