missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAST2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558900
This item is not returnable.
View return policy
Description
Immunogen sequence: GESGEEDPFP SRDPRSLGPM VPSLLTGITL GPPRMESPSG PHRRLGSPQA IEEAASSSSA GPNLGQSG Highest antigen sequence identity to the following orthologs: Mouse - 46%, Rat - 46%.
Syntrophin is an adapter protein that functions to bind certain signaling molecules to the dystrophin-associated protein complex. This complex connects the extracellular matrix to the intracellular cytoskeleton for construction and maintenance of the postsynaptic structures in the neuromuscular junction and the central nervous system. Microtubule-associated serine/threonine-protein kinase 2 (MAST205) is a testis-specific, cytoplasmic protein that functions in a multi-protein complex in the maturation of spermatids. MAST205 is involved in linking the dystrophin/utrophin network with microtubule filaments via Syntrophin. By forming a complex with TRAF6, MAST205 regulates lipopolysaccharide-induced IL-12 synthesis in macrophages. This leads to the inhibition of TRAF6 NF-kappa-B activation. Two isoforms exist for MAST205 due to alternative splicing. Isoform 1 represents the full length protein, while isoform 2 lacks the residues 327-396 and 1091-1113. The N-terminus of MAST205 must be phosphorylated in order for ubiquitination to occur at the same site. This ubiquitination leads to the degradation of MAST205 via proteasome-mediated proteolysis.
Specifications
| MAST2 | |
| Polyclonal | |
| Unconjugated | |
| MAST2 | |
| KIAA0807; Mast2; MAST205; microtubule associated serine/threonine kinase 2; microtubule associated testis specific serine/threonine protein kinase; microtubule-associated serine/threonine-protein kinase 2; Mtssk | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23139 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6P0Q8 | |
| MAST2 | |
| Recombinant protein corresponding to Human MAST2. Recombinant protein control fragment (Product #RP-98307). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction