missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mast Cell Marker Antibody (MCG35) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£159.00 - £307.00
Specifications
| Antigen | Mast Cell Marker |
|---|---|
| Clone | MCG35 |
| Concentration | 1 mg/mL |
| Dilution | Immunohistochemistry 1:10 - 1:500 |
| Applications | Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18674666
|
Novus Biologicals
NBP2-50484-0.025ml |
0.025 mL |
£159.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18617528
|
Novus Biologicals
NBP2-50484 |
0.1 mL |
£307.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mast Cell Marker Monoclonal antibody specifically detects Mast Cell Marker in Human samples. It is validated for ImmunohistochemistrySpecifications
| Mast Cell Marker | |
| 1 mg/mL | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Cardiovascular Biology | |
| PBS | |
| IgG1 κ | |
| Protein A purified |
| MCG35 | |
| Immunohistochemistry 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Spleen cells and bone marrow cells (erythrocyte depleted) from a patient with systemic mastocytosis. The bone marrow preparation consisted of 50% typical mast cells. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title