missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mast Cell Chymase Recombinant Rabbit Monoclonal Antibody (23GB5095), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | Mast Cell Chymase |
|---|---|
| Clone | 23GB5095 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30088522
|
Invitrogen™
MA552788 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088134
|
Invitrogen™
MA552789 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mast Cell Chymase Recombinant Monoclonal antibody specifically detects Mast Cell Chymase in Mouse samples. It is validated for Western BlotSpecifications
| Mast Cell Chymase | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| P21844 | |
| 17228 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5095 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| alpha-chymase; Chymase; chymase 1; chymase 1 preproprotein transcript E; chymase 1 preproprotein transcript I; chymase 1, mast cell; chymase, heart; chymase, mast cell; CMA1; CYH; CYM; mast cell; Mast cell chymase 1; mast cell protease 3; Mast cell protease 5; mast cell protease I; Mast cell protease III; MCP3P; Mcp5; Mcp-5; Mcpt3; Mcpt5; MCT1; mMCP-5; rMCP-3; rMCP-5; rMCP-III | |
| A synthesized peptide derived from human Mast Cell Chymase (204-247AA). | |
| Primary | |
| -20°C | |
| CMA1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title