missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MassRuler DNA Ladders, ready-to-use
GREENER_CHOICE

Product Code. 10849540 Shop All Thermo Scientific Products
Change view
Click to view available options
Size Range:
100 to 1000 bp
1500 to 10000 bp
80 to 10000 bp
80 to 1031 bp
Unit Size:
Pack of 2
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Range unitSize
10849540 80 to 1031 bp Pack of 2
10170690 80 to 10000 bp Pack of 2
10736041 100 to 1000 bp Pack of 2
10746231 100 to 1000 bp Pack of 2
10374380 1500 to 10000 bp Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10849540 Supplier Thermo Scientific™ Supplier No. SM0383
Limited-Time Offers on Selected Molecular Biology Essentials
Use promo code 27945
View all promotions
 Restricted Item
Estimated Shipment: 17-07-2026
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Thermo Scientific MassRuler DNA Ladders are designed for quantification and sizing of DNA fragments by agarose gel electrophoresis.

Thermo Scientific MassRuler DNA Ladders are designed for accurate quantification and sizing of DNA fragments by agarose gel electrophoresis. Individuals DNA fragments are chromatography-purified and calibrated against a standard that guarantees precise quantity of each band.

Highlights

  • Accurate DNA sizing and quantification – DNA ladder fragments are quantified spectrophotometrically
  • Sharp bands, easy-to-remember fragment sizes and quantities
  • Ready-to-use—premixed with 6X MassRuler DNA Loading Dye for direct loading
  • Stable at room temperature for up to 24 months

Sustainability features

Ready-to-use MassRuler DNA ladders can be stored at room temperature for up to 24 months. The ladder is shipped at ambient temperature, using optimized right-sized packaging and 100% recyclable packaging reducing waste.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 6X
Gel Type Agarose
Label or Dye Bromophenol Blue
Ready to Load Yes
Size Range 80 to 1031 bp
Gel Compatibility Agarose Gels
Quantity 2 x 500 μL
Product Line MassRuler
Product Type DNA Ladder

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.