Learn More
PeproTech Maspin Polyclonal Antibody, PeproTech®, Invitrogen™

Rabbit Polyclonal Antibody
Brand: PeproTech 500-P270-1MG
Description
AA Sequence of recombinant protein: MDALQLANSA FAVDLFKQLC EKEPLGNVLF SPICLSTSLS LAQVGAKGDT ANEIGQVLHF ENVKDIPFGF QTVTSDVNKL SSFYSLKLIK RLYVDKSLNL STEFISSTKR PYAKELETVD FKDKLEETKG QINNSIKDLT DGHFENILAD NSVNDQTKIL VVNAAYFVGK WMKKFPESET KECPFRLNKT DTKPVQMMNM EATFCMGNID SINCKIIELP FQNKHLSMFI LLPKDVEDES TGLEKIEKQL NSESLSQWTN PSTMANAKVK LSIPKFKVEK MIDPKACLEN LGLKHIFSED TSDFSGMSET KGVALSNVIH KVCLEITEDG GDSIEVPGAR ILQHKDELNA DHPFIYIIRH NKTRNIIFFG KFCSP. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human Maspin. Anti-Human Maspin-specific antibody was purified by affinity chromatography employing an immobilized Human Maspin matrix. Sandwich ELISA: To detect Human Maspin by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human Maspin (500-P270Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human Maspin. Western Blot: To detect Human Maspin by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human Maspin is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
SERPINB5 is a tumor suppressor. It blocks the growth, invasion, and metastatic properties of mammary tumors. As it does not undergo the S (stressed) to R (relaxed) conformational transition characteristic of active serpins, it exhibits no serine protease inhibitory activity.
Specifications
| Maspin | |
| Polyclonal | |
| Unconjugated | |
| SERPINB5 | |
| 1110036M19Rik; AI462524; AI646751; Maspin; ovalbumin; peptidase inhibitor 5; PI5; PI-5; protease inhibitor 5; protease inhibitor 5 (maspin); serine (or cysteine) peptidase inhibitor, clade B, member 5; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 5; serine (or cysteine) proteinase inhibitor, clade B, member 5; serine protease inhibitor 7; serpin B5; serpin family B member 5; serpin peptidase inhibitor, clade B (ovalbumin), member 5; SERPINB5; Spi5; Spi7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5268 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| P36952 | |
| SERPINB5 | |
| E.coli-derived Recombinant Human Maspin. | |
| 1 mg | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.