missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Maspin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA527382
This item is not returnable.
View return policy
Description
Recommended positive controls: A431, HeLaS3, HepG2. Predicted reactivity: Mouse (85%), Rat (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
SERPINB5 is a tumor suppressor. It blocks the growth, invasion, and metastatic properties of mammary tumors. As it does not undergo the S (stressed) to R (relaxed) conformational transition characteristic of active serpins, it exhibits no serine protease inhibitory activity.
Specifications
| Maspin | |
| Polyclonal | |
| Unconjugated | |
| SERPINB5 | |
| 1110036M19Rik; AI462524; AI646751; Maspin; ovalbumin; peptidase inhibitor 5; PI5; PI-5; protease inhibitor 5; protease inhibitor 5 (maspin); serine (or cysteine) peptidase inhibitor, clade B, member 5; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 5; serine (or cysteine) proteinase inhibitor, clade B, member 5; serine protease inhibitor 7; serpin B5; serpin family B member 5; serpin peptidase inhibitor, clade B (ovalbumin), member 5; SERPINB5; Spi5; Spi7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5268 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.81 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P36952 | |
| SERPINB5 | |
| Recombinant protein encompassing a sequence within the center region of human MASPIN. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction