Learn More
Maspin Mouse anti-Human, Unconjugated, Clone: 3G5A3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67633-1-IG-150UL
This item is not returnable.
View return policy
Description
The Maspin antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Maspin. This antibody recognizes human antigen. The Maspin antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| Maspin | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, Immunohistochemistry, ELISA | |
| 3G5A3 | |
| Unconjugated | |
| 150 μL | |
| 5268 | |
| IgG2a |