missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MASP2 Antibody (narsoplimab) [Janelia Fluor™ 635], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28061JF635
This item is not returnable.
View return policy
Description
MASP2 Monoclonal antibody specifically detects MASP2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| MASP2 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| EC 3.4.21, EC 3.4.21.104, mannan-binding lectin serine peptidase 1 pseudogene 1, mannan-binding lectin serine peptidase 2, mannan-binding lectin serine protease 1 pseudogene 1, mannan-binding lectin serine protease 2, Mannose-binding protein-associated serine protease 2, MAP19, MASP1P1, MASP-2, MBL-associated plasma protein of 19 kD, MBL-associated serine protease 2, small MBL-associated protein, sMAP | |
| MASP2 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| narsoplimab | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| 10747 | |
| Store at 4°C in the dark. | |
| IgG4 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction