missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MASP1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

£133.00 - £430.00

Specifications

Antigen MASP1
Concentration 0.73 mg/mL
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17750674
GREENER_CHOICE
View Documents
Invitrogen™
PA5119268
200 μL
£430.00
200µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17703184
View Documents
Invitrogen™
PA5121782
20 μL
£133.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MASP1 Polyclonal Antibody for IHC (P), Western Blot

MBL is a key component in immune response in that it can directly trigger neutralization of invading microorganisms by an Ab-independent mechanism. Mutations of human MBL are associated with immunodeficiency resulting from a reduction in the ability of the mutant MBL to initiate complement fixation. In human, three types of MBL-associated serine proteases, MASP-1, MASP-2 and MASP-3, and a truncated form of MASP-2 (small MBL-associated protein; sMAP or MAp19) complex with MBL to activate the lectin pathway of the complement system. MASP-3 is an alternatively spliced product from the MASP-1 gene. The heavy/A chains are identical between MASP-1 and MASP-3 but the light/B chains are entirely different. Activated MASPs subsequently cleave and activate downstream components of the complement pathway.
Specifications

Specifications

MASP1
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Polyclonal
Liquid
IgG
Human, Mouse
MASP1
3MC1; AW048060; C4/C2 activating component of Ra-reactive factor; CCPII; Complement factor MASP-3; complement-activating component of Ra-reactive factor; Crarf; CRARF1; mannan binding lectin serine peptidase 1; mannan-binding; mannan-binding lectin serine peptidase 1; mannan-binding lectin serine peptidase 1 (C4/C2 activating component of Ra-reactive factor); mannan-binding lectin serine protease 1; Mannan-binding lectin serine protease 1 heavy chain; Mannan-binding lectin serine protease 1 light chain; Mannose-binding lectin-associated serine protease 1; mannose-binding protein associated serine protease-1; mannose-binding protein-associated serine protease; MAP1; MAp44; MASP; MASP1; MASP-1; Masp1/3; MASP3; PRSS5; Ra-reactive factor serine protease p100; raRF; Serine protease 5; serine protease MASP1
MASP1
Primary
Antigen affinity chromatography, Protein A
0.73 mg/mL
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
P48740, P98064
17174, 5648
Recombinant protein Mannose Associated Serine Protease 1. The antigen corresponds to amino acid range 20-447 of the target protein.
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.