missing translation for 'onlineSavingsMsg'
Learn More

Maslinic Acid, TRC

Product Code. 30005705
Change view
Click to view available options
Quantity:
5 mg
50 mg
100 mg
Unit Size:
100mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005705 5 mg 5mg
30010331 50 mg 50mg
30011863 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005705 Supplier Toronto Research Chemicals Supplier No. TRCM1975505MG
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4373-41-5
Molecular Formula C30 H48 O4
Molecular Weight (g/mol) 472.7
Synonym Olean-12-en-28-oic acid, 2,3-dihydroxy-, (2α,3β)-, Maslinic acid (6CI), Olean-12-en-28-oic acid, 2α,3β-dihydroxy- (7CI,8CI), (2α,3β)-2,3-Dihydroxyolean-12-en-28-oic acid, 2α,3β-Dihydroxyloleanolic acid, 2α,3β-Dihydroxyolean-12-en-28-oic acid, 2α-Hydroxyoleanolic acid, Crataegolic acid, Crategolic acid, Masilinic acid, Maslic acid
IUPAC Name (4aS,6aR,6aS,6bR,8aR,10R,11R,12aR,14bS)-10,11-dihydroxy-2,2,6a,6b,9,9,12a-heptamethyl-1,3,4,5,6,6a,7,8,8a,10,11,12,13,14b-tetradecahydropicene-4a-carboxylic acid
SMILES CC1(C)CC[C@@]2(CC[C@]3(C)C(=CC[C@@H]4[C@@]5(C)C[C@@H](O)[C@H](O)C(C)(C)[C@@H]5CC[C@@]34C)[C@@H]2C1)C(=O)O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C30H48O4/c1-25(2)12-14-30(24(33)34)15-13-28(6)18(19(30)16-25)8-9-22-27(5)17-20(31)23(32)26(3,4)21(27)10-11-29(22,28)7/h8,19-23,31-32H,9-17H2,1-7H3,(H,33,34)/t19-,20+,21-,22+,23-,27-,28+,29+,30-/m0/s1
Formula Weight 472.3553
Percent Purity >95%
Chemical Name or Material Crategolic Acid (Maslinic Acid)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.