missing translation for 'onlineSavingsMsg'
Learn More

Maslinic acid, MedChemExpress

Product Code. 16453886
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16453886 25 mg 25mg
16449048 10 mg 10mg
16421468 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16453886 Supplier Medchem Express Supplier No. HYN0629
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Maslinic acid can inhibit the DNA-binding activity of NF-κB p65 and abolish the phosphorylation of IκB-α, which is required for p65 activation.

Maslinic acid can inhibit the DNA-binding activity of NF-κB p65 and abolish the phosphorylation of IκB-α, which is required for p65 activation.

Specifications

Chemical Name or Material Maslinic acid
CAS 4373-41-5
Color White
Purity Grade Notes Research
Molecular Formula C30H48O4
Quantity 25 mg
Synonym Crategolic acid 2α-Hydroxyoleanolic acid
Solubility Information DMSO : 100 mg/mL (211.55 mM; Need ultrasonic)
SMILES C[C@]([C@@]1([H])[C@]2(C[C@@H](O)[C@@H]3O)C)(CC[C@@]2([H])C3(C)C)[C@@](C4=CC1)(CC[C@]5(C(O)=O)[C@@]4([H])CC(C)(C)CC5)C
Molecular Weight (g/mol) 472.7
Formula Weight 472.7
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.