Learn More
MASH1 Monoclonal Antibody (24B72D11), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£132.00 - £317.00
Specifications
Antigen | MASH1 |
---|---|
Clone | 24B72D11 |
Concentration | 0.5 mg/mL |
Applications | Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15547456
|
Affymetrix eBioscience
14-5794-80 |
25 μg |
£132.00
25µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15557456
|
Affymetrix eBioscience
14-5794-82 |
100 μg |
£317.00
100µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The monoclonal antibody 24B72D11 recognizes mouse and rat Mammalian achaete-scute homolog 1 (MASH1), a 34 kDa basic helix-loop-helix (bHLH) transcription factor. MASH1, like family members Olig2 and Ngn2, is expressed in progenitor cells throughout the nervous system and retina and is a key regulator of cell fate specification. Expression of MASH1 is regulated by the Delta/Notch1 pathway through direct binding of Hes1 to MASH1 as well as competitive binding to MASH1 hetero-dimerizing proteins. At a transcriptional level, MASH1 expression is controlled by notch-mediated regulation of sox2, which binds to the MASH1 promoter region up-regulating expression. MASH1 has been implicated in the formation of multiple neural cell types including dopaminergic, serotonergic, glutaminergic and GABAergic neurons. Overexpression of MASH1 has been shown in small cell lung carcinoma and medullary thyroid cancer. Applications Reported: This 24B72D11 antibody has been reported for use in western blotting, immunohistochemical staining of frozen tissue sections, and immunocytochemistry. Applications Tested: This 24B72D11 antibody has been tested by western blot on reduced PC12 cells and can be used at less than or equal to 5 μg/mL. The 24B72D11 antibody has also been tested by immunocytochemistry on fixed and permeablized PC12 cells at can be used at less than or equal to 20 μg/mL.
ASCL1 (also known as ASH1) is a basic helix-loop-helix transcription factor that is required for early development of the nervous system. Expressed in fetal brain, ASCL1 is essential for the proper development of autonomic neurons and for the survival of subsets of autonomic neurons. ASCL1 interaction with MEF-2A may regulate the expression of specific genes that are critical for the formation of distinct neuronal circuits within the central nervous system. The high level of ASCL1 expression in neuroendocrine tumors, such as medullary thyroid cancer, small cell lung cancer and lung cancer with neuroendocrine features may provide a useful marker for cancers with neuroendocrine features. Mapping to human chromosome 12, the ASCL1 gene contains a trinucleotide repeat region, making this locus a candidate for inherited disease.Specifications
MASH1 | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
achaete scute protein; achaete-scute complex homolog 1; achaete-scute complex homolog 1 (Drosophila); achaete-scute complex homolog-like 1; achaete-scute complex-like 1; achaete-scute complex-like protein 1; achaete-scute family bHLH transcription factor 1; achaete-scute homolog 1; AI225900; Ascl1; ASH1; ASH-1; bHLHa46; Class A basic helix-loop-helix protein 46; HASH1; KMT2H; mammalian achaete scute homolog 1; Mash1; mASH-1 | |
Ascl1 | |
Primary | |
4° C | |
Ascl1 |
24B72D11 | |
Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot | |
Unconjugated | |
Mouse | |
Mouse, Rat | |
P19359, Q02067 | |
17172, 64186 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.