missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MAS1L Monoclonal antibody specifically detects MAS1L in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
Specifications
| Antigen | MAS1L |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | SR1619 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 1:50 |
| Formulation | PBS, pH 7.4, 150mM NaCl, 50% glycerol. |
| Gene Alias | dJ994E9.2, MAS1 oncogene-like, MAS1-like, MAS-L, MAS-R, mas-related G protein-coupled MRG, mas-related G-protein coupled receptor MRG, MRGMGC119987 |
| Host Species | Rabbit |
| Immunogen | A synthesized peptide derived from human MAS1L (Uniprot #: P35410) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?