missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533918
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey, Mouse, Rat, Bovine, Dolphin, Hamster, Elephant, Panda, Pig, Opossum (100%) Platypus (94%).
The structure of the MAS1 product indicates that it belongs to the class of receptors that are coupled to GTP-binding proteins and share a conserved structural motif, which is described as a 7-transmembrane segment following the prediction that these hydrophobic segments form membrane-spanning alpha-helices. The MAS1 protein may be a receptor that, when activated, modulates a critical component in a growth-regulating pathway to bring about oncogenic effects.
Specifications
| MAS1 | |
| Polyclonal | |
| Unconjugated | |
| MAS1 | |
| angiotensin-(1-7) receptor; c-mas; MAS; MAS proto-oncogene; MAS1; Mas-1; MAS1 oncogene; Mas1 proto-oncogene; MAS1 proto-oncogene, G protein-coupled receptor; Mas-related G protein-coupled receptor a; MGC119966; MGRA; Proto-oncogene Mas | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100522470, 17171, 25153, 4142, 783010 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| PBS with 0.1% sodium azide | |
| P04201, P12526, P30554 | |
| MAS1 | |
| Synthetic 16 amino acid peptide from 1st cytoplasmic domain of human MAS1. | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat, Monkey, Bovine, Hamster, Pig | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction