missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MAS™ PAR™ TDM Controls

Product Code. 17476883 Shop All Thermo Scientific Products
Change view
Click to view available options
Level:
1
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Level Unit Size
17476883 1 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 17476883 Supplier Thermo Scientific™ Supplier No. PTD11001
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Check therapeutic drug monitoring test procedures with ready-to-use, liquid assayed multi-analyte MAS PAR TDM Controls.

Check therapeutic drug monitoring test procedures with ready-to-use, liquid assayed multi-analyte Thermo Scientific™ MAS™ PAR™ TDM Controls, therapeutic drug controls with assayed values designed for therapeutic drugs and common immunoassayed test procedures on most automated instruments.

  • Liquid, ready to use
  • Features three significant levels
  • Includes values for major automated analyzers
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description MAS PAR TDM Quality Control
Detectable Analytes Acetaminophen, Amikacin, Caffeine, Carbamazepine, Digoxin, Disopyramide, Estriol, Ethanol, Ethosuximide, Gentamicin, Lidocaine, Lithium, Methotrexate, N-acetylprocainamide (NAPA), Phenobarbital, Phenytoin, Primidone, Procainamide, Quinidine, Salicylate (SALY), Theophylline, Thyroid Stimulating Hormone (TSH), Thyroxine (T4 Total), Tobramycin, Tricyclic Antidepressants, Triiodthyronine Total (TT3), Valproic Acid, Vancomycin
Sample Type Serum
Type Control
Level 1
Quantity 6 x 5 mL
Stability 36 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.