missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MAS™ Omni·IMMUNE™ Immunoassay Controls

Product Code. p-4527717 Shop All Thermo Scientific Products
Click to view available options
Level:
1
Unit Size:
Pack of 6
This item is not returnable. View return policy

Product Code. 16594704

Brand: Thermo Scientific™ OIM101

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Consolidate routine immunoassay, tumor marker and specialty immunoassay QC processing into a single product offering three distinct levels.

Consolidate routine immunoassay, tumor marker and specialty immunoassay QC processing into a single product with Thermo Scientific™ MAS™ Omni·IMMUNE Immunoassay Controls.

  • Liquid, ready-to-use
  • Human serum based matrix
  • Value assignment provided for most major analyzer
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -25°C to -15°C
Description Omni IMMUNE Control, Assayed
Detectable Analytes 17-OH-Progesterone, 25-OH-Vitamin D, Acetaminophen, Adrenocorticotropic Hormone (ACTH), Aldosterone, alpha-Fetoprotein (AFP), Amikacin, Benzodiazepines, beta-2-microglobulin (B2M), β-hCG, CA-125, CA 15-3, CA 19-9, Carbamazepine, Carcinoembryonic Antigen (CEA), Cortisol, C-peptide, DHEA Sulfate, Digoxin, Disopyramide, Estradiol, Estriol (Free), Ethosuximide, Ferritin, Folate (Vitamin B9), Fructosamine, Follicle Stimulating Hormone (FSH), Gastrin, Gentamicin, Growth Hormone, hCG, Homocysteine, IgE, Insulin-like Growth Factor 1 (IGF-1), Insulin, Luteinizing Hormone (LH), Lidocaine, Lithium, N-acetylprocainamide (NAPA), Prostatic Acid Phosphatase (PAP), Phenobarbital, Phenytoin, Phenytoin (Free), Primidone, Procainamide, Procalcitonin (PCT), Progesterone, Prolactin, PSA (Free), PSA (Total), Parathyroid Hormone (PTH), Quinidine, Salicylate (SALY), T3 (Free), Triiodothyroxine (T3 Total), T4 (Free), Thyroxine (T4 Total), Thyroxine-binding Globulin (TBG), Testosterone, Theophylline, Thyroglobulin, Tobramycin, Tricyclic Antidepressants, Thyroid Stimulating Hormone (TSH), T-uptake, Valproic Acid, Valproic Acid (Free), Vancomycin, Vitamin B12
Sample Type Serum
Type Control
Level 1
Packaging 6 Pk.
Quantity 5 mL
Stability 2° to 8°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.