missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MAS™ Omni·IMMUNE™ Immunoassay Controls

Product Code. 16594704 Shop All Thermo Scientific Products
Change view
Click to view available options
Level:
1
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Level Unit Size
16594704 1 Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16594704 Supplier Thermo Scientific™ Supplier No. OIM101
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Consolidate routine immunoassay, tumor marker and specialty immunoassay QC processing into a single product offering three distinct levels.

Consolidate routine immunoassay, tumor marker and specialty immunoassay QC processing into a single product with Thermo Scientific™ MAS™ Omni·IMMUNE Immunoassay Controls.

  • Liquid, ready-to-use
  • Human serum based matrix
  • Value assignment provided for most major analyzer
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -25°C to -15°C
Description Omni IMMUNE Control, Assayed
Detectable Analytes 17-OH-Progesterone, 25-OH-Vitamin D, Acetaminophen, Adrenocorticotropic Hormone (ACTH), Aldosterone, alpha-Fetoprotein (AFP), Amikacin, Benzodiazepines, beta-2-microglobulin (B2M), β-hCG, CA-125, CA 15-3, CA 19-9, Carbamazepine, Carcinoembryonic Antigen (CEA), Cortisol, C-peptide, DHEA Sulfate, Digoxin, Disopyramide, Estradiol, Estriol (Free), Ethosuximide, Ferritin, Folate (Vitamin B9), Fructosamine, Follicle Stimulating Hormone (FSH), Gastrin, Gentamicin, Growth Hormone, hCG, Homocysteine, IgE, Insulin-like Growth Factor 1 (IGF-1), Insulin, Luteinizing Hormone (LH), Lidocaine, Lithium, N-acetylprocainamide (NAPA), Prostatic Acid Phosphatase (PAP), Phenobarbital, Phenytoin, Phenytoin (Free), Primidone, Procainamide, Procalcitonin (PCT), Progesterone, Prolactin, PSA (Free), PSA (Total), Parathyroid Hormone (PTH), Quinidine, Salicylate (SALY), T3 (Free), Triiodothyroxine (T3 Total), T4 (Free), Thyroxine (T4 Total), Thyroxine-binding Globulin (TBG), Testosterone, Theophylline, Thyroglobulin, Tobramycin, Tricyclic Antidepressants, Thyroid Stimulating Hormone (TSH), T-uptake, Valproic Acid, Valproic Acid (Free), Vancomycin, Vitamin B12
Sample Type Serum
Type Control
Level 1
Packaging 6 Pk.
Quantity 5 mL
Stability 2° to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.