missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MAS™ Bilirubin Controls

Product Code. 17274296 Shop All Thermo Scientific Products
Change view
Click to view available options
Level:
3
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Level Unit Size
17274296 3 Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17274296 Supplier Thermo Scientific™ Supplier No. BC103
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Monitor the performance of total, conjugated, unconjugated and neonatal Bilirubin with tri-level, assayed MAS Bilirubin Controls.

Monitor the performance of total, conjugated, unconjugated and neonatal Bilirubin with tri-level, assayed Thermo Scientific™ MAS™ Bilirubin Controls. These controls are also available in a convenient Vista™ vial format.

  • Liquid, ready-to-use; no additional preparation steps required
  • Bovine serum matrix
  • Assayed values available for most major analyzers
  • Elevated bilirubin in Level 3 for neonatal applications
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Two year shelf life from date of manufacture at 2° to 8°C; 14 days open vial at 2° to 8°C when stored in the dark
Description Bilirubin Control, Assayed
Detectable Analytes Bilirubin Conjugated, Bilirubin Total (TBIL), Bilirubin Direct (DBIL), Bilirubin Unconjugated, Bilirubin Neonatal
Sample Type Serum
Type Control
Level 3
Packaging 6 Pk.
Quantity 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.