missing translation for 'onlineSavingsMsg'
Learn More

Mas Antibody, DyLight 405, Novus Biologicals™

Product Code. 18394745 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mL
Unit Size:
0.05mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18394745 0.05 mL 0.05mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18394745 Supplier Novus Biologicals Supplier No. NLS1531V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Mas Polyclonal specifically detects Mas in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mas
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate DyLight 405
Dilution Flow Cytometry, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:500, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. P04201
Gene Alias MAS, MAS1 oncogene, MGC119966, proto-oncogene Mas
Gene Symbols MAS1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide made to the C-terminus of human MAS1. [UniProt# P04201]
Purification Method Affinity Purified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Cancer, GPCR, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 4142
Test Specificity Human and mouse. Immunogen sequence has 82% identity with rat.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.