missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mas Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-78444B
This item is not returnable.
View return policy
Description
Mas Polyclonal specifically detects Mas in Human, Mouse, Rat, Primate, Rabbit samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Mas | |
| Polyclonal | |
| Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| P04201 | |
| MAS1 | |
| A synthetic peptide made to an internal portion of the human MAS1 protein (between residues 75-125) [Uniprot: P04201] | |
| 0.1 mL | |
| Cancer, GPCR, Neuroscience | |
| 4142 | |
| Human, Mouse, Rat, Primate, Rabbit | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| MAS, MAS1 oncogene, MGC119966, proto-oncogene Mas | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, monkey, rat, and mouse. Rabbit reactivity reported in the scientific literature (PMID: 23701246). | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction