missing translation for 'onlineSavingsMsg'
Learn More

Maryland Pesticide Mixture 1 1000 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30310193
Change view
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30310193 5 x 1 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30310193 Supplier Dr. Ehrenstorfer Supplier No. DRES50000140AL
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (E)-Fenpyroximate∣Abamectin∣Acetamiprid∣Aldicarb∣Ancymidol∣Azoxystrobin∣Carbaryl∣Carbofuran∣Chlorantraniliprole∣Dimethoate∣Ethephon∣Etoxazole∣Flonicamid∣Fludioxonil∣Imidacloprid∣Methomyl∣Myclobutanil∣Propiconazole∣Thiacloprid∣Thiamethoxam
CAS 134098-61-6, 71751-41-2, 135410-20-7, 116-06-3, 12771-68-5, 131860-33-8
Analyte or Component Names (E)-Fenpyroximate∣Abamectin∣Acetamiprid∣Aldicarb∣Ancymidol∣Azoxystrobin∣Carbaryl∣Carbofuran∣Chlorantraniliprole∣Dimethoate∣Ethephon∣Etoxazole∣Flonicamid∣Fludioxonil∣Imidacloprid∣Methomyl∣Myclobutanil∣Propiconazole∣Thiacloprid∣Thiamethoxam
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Acetonitrile
Industry Type Food Testing
Quantity 5 x 1 mL
Concentration or Composition (by Analyte or Components) (E)-Fenpyroximate 1000 μg/mL∣Abamectin 1000 μg/mL∣Acetamiprid 1000 μg/mL∣Aldicarb 1000 μg/mL∣Ancymidol 1000 μg/mL∣Azoxystrobin 1000 μg/mL∣Carbaryl 1000 μg/mL∣Carbofuran 1000 μg/mL∣Chlorantraniliprole 1000 μg/mL∣Dimethoate 1000 μg/mL∣Ethephon 1000 μg/mL∣Etoxazole 1000 μg/mL∣Flonicamid 1000 μg/mL∣Fludioxonil 1000 μg/mL∣Imidacloprid 1000 μg/mL∣Methomyl 1000 μg/mL∣Myclobutanil 1000 μg/mL∣Propiconazole 1000 μg/mL∣Thiacloprid 1000 μg/mL∣Thiamethoxam 1000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.