missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MARS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14829-1-AP-20UL
This item is not returnable.
View return policy
Description
MARS is a member of aminoacyl-tRNA synthetases which are a class of enzymes that charge tRNAs with their cognate amino acids. The protein encoded by this gene belongs to the class I family of tRNA synthetases.Specifications
| MARS | |
| Polyclonal | |
| Unconjugated | |
| MARS | |
| MARS, Methionine tRNA ligase, methionyl tRNA synthetase, METRS, MTRNS | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 216443, 299851, 4141 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Western Blot, Immunocytochemistry, Immunoprecipitation | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P56192, Q68FL6 | |
| MARS | |
| MARS Fusion Protein Ag6619 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction