missing translation for 'onlineSavingsMsg'
Learn More

Maropitant, MedChemExpress

Product Code. 16483068
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
2mg
5mg
This item is not returnable. View return policy

Product Code. 16483068

Brand: Medchem Express HY10053

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Maropitant is a selective and orally active neurokinin (NK1) receptor antagonist. Maropitant acts by blocking the binding of substance P within the emetic center and the chemoreceptor trigger zone (CRTZ). Maropitant is highly effective in preventing vomiting.

Maropitant is a selective and orally active neurokinin (NK1) receptor antagonist. Maropitant acts by blocking the binding of substance P within the emetic center and the chemoreceptor trigger zone (CRTZ). Maropitant is highly effective in preventing vomiting.

Specifications

Chemical Name or Material Maropitant
CAS 147116-67-4
Color Pink
Purity Grade Notes Research
Molecular Formula C32H40N2O
Quantity 10 mg
Solubility Information DMSO : 33.33 mg/mL (71.12 mM; Need ultrasonic)
SMILES CC(C)(C)C1=CC=C(OC)C(CN[C@@H]2[C@H](C(C3=CC=CC=C3)C4=CC=CC=C4)N5CCC2CC5)=C1
Molecular Weight (g/mol) 468.67
Formula Weight 468.67
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.