missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MARK4 Recombinant Human Protein
MARK4 is a serine/threonine protein kinase.
£2555.00 - £12060.00
Specifications
Accession Number | NP_113605 |
---|---|
For Use With (Application) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Formulation | 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol |
Molecular Weight (g/mol) | 102.8kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15438533
|
Invitrogen™
PR7177C |
100μg |
£2555.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10066932
|
Invitrogen™
PV4175 |
1mg |
£12060.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MARK4 is likely to be directly involved in microtubule organization in neuronal cells.
Specifications
NP_113605 | |
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
CAMKL Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
102.8kDa | |
RUO - research use only | |
MARK4 | |
Human | |
GST Tag | |
Frozen |