missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MARK4 Monoclonal Antibody (OTI9B7)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527002
This item is not returnable.
View return policy
Description
MARK4 Monoclonal Antibody for Western Blot, IHC (P)
MARK4 is a serine/threonine kinase that is likely to be directly involved in microtubule organization in neuronal cells. The encoded protein is associated with the centrosome throughout mitosis and may be involved in cell cycle control. Expression of MARK4 is a potential marker for cancer, and the encoded protein may also play a role in Alzheimer's disease. Pseudogenes of MARK4 are located on both the short and long arm of chromosome 3. Alternatively spliced transcript variants encoding multiple isoforms have been observed for MARK4.
Specifications
| MARK4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96L34 | |
| Mark4 | |
| Human recombinant protein fragment corresponding to amino acids 390-467 of MARK4 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI9B7 | |
| Unconjugated | |
| Mark4 | |
| 2410090P21Rik; C79806; FLJ90097; KIAA1860; MAP/microtubule affinity regulating kinase 4; MAP/microtubule affinity-regulating kinase 4; MAP/microtubule affinity-regulating kinase 4L; MAP/microtubule affinity-regulating kinase like 1; MAP/microtubule affinity-regulating kinase-like 1; Mark4; MARK4 serine/threonine protein kinase; MARK4L; MARK4S; MARKL1; MARKL1L; microtubule affinity regulating kinase 4; Nbla00650; PAR-1D | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 57787 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction