missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MARK4 Antibody (OTI9B7), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72591AF647
This item is not returnable.
View return policy
Description
MARK4 Monoclonal specifically detects MARK4 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.
Specifications
| MARK4 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MARK4 | |
| Recombinant protein fragment corresponding to amino acids 390-467 of human MARK4 (NP_113605) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Immunohistochemistry | |
| OTI9B7 | |
| Immunohistochemistry | |
| EC 2.7.11, EC 2.7.11.1, FLJ12177, FLJ90097, KIAA1860MARK4L, MAP/microtubule affinity-regulating kinase 4, MAP/microtubule affinity-regulating kinase like 1, MAP/microtubule affinity-regulating kinase-like 1, MARK4 serine/threonine protein kinase, MARKL1L, MARKL1MARK4S, Nbla00650 | |
| Mouse | |
| Affinity Purified | |
| Apoptosis | |
| 57787.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction