missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MARK3 Recombinant Rabbit Monoclonal Antibody (JE44-84)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA534841
This item is not returnable.
View return policy
Description
Positive Control: Rat brain tissue lysate, A431, 293, MCF-7, SH-SY-5Y, SiHa, human appendix tissue, human breast cancer tissue, mouse small intestine tissue.
MARK3 is a member of a group of kinases known to regulate cell polarity via phosphorylation of microtubule-associated proteins (MAPs). The protein encoded by MARK3 is activated by phosphorylation and in turn is involved in the phosphorylation of tau proteins MAP2 and MAP4. Several transcript variants encoding different isoforms have been found for MARK3.
Specifications
| MARK3 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P27448 | |
| MARK3 | |
| Synthetic peptide within Human MARK3 aa 567-616. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| JE44-84 | |
| Unconjugated | |
| MARK3 | |
| 1600015G02Rik; A430080F22Rik; cdc25C-associated protein kinase 1; cTAK1; C-TAK1; ELKL motif kinase 2; Emk2; EMK-2; EMK2/MARK3 serine-threonine protein kinase; ETK1; ETK-1; KP78; MAP/microtubule affinity regulating kinase 3; MAP/microtubule affinity-regulating kinase 3; MARK3; microtubule affinity regulating kinase 3; Mpk10; MPK-10; PAR1A; Par-1a; Protein kinase STK10; ser/Thr protein kinase PAR-1; Serine/threonine-protein kinase p78 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 4140 | |
| -20°C, Avoid Freeze/Thaw Cycles, store in dark | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction