Learn More
Invitrogen™ MARK3 Recombinant Rabbit Monoclonal Antibody (24GB3795)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557063
 View more versions of this product
Product Code. 30275122
  • £434.00 / 100µL
Description
MARK3 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
MARK3 is a member of a group of kinases known to regulate cell polarity via phosphorylation of microtubule-associated proteins (MAPs). The protein encoded by MARK3 is activated by phosphorylation and in turn is involved in the phosphorylation of tau proteins MAP2 and MAP4. Several transcript variants encoding different isoforms have been found for MARK3.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MARK3
Recombinant Monoclonal
Unconjugated
MARK3
1600015G02Rik; A430080F22Rik; cdc25C-associated protein kinase 1; cTAK1; C-TAK1; ELKL motif kinase 2; Emk2; EMK-2; EMK2/MARK3 serine-threonine protein kinase; ETK1; ETK-1; KP78; MAP/microtubule affinity regulating kinase 3; MAP/microtubule affinity-regulating kinase 3; MARK3; microtubule affinity regulating kinase 3; Mpk10; MPK-10; PAR1A; Par-1a; Protein kinase STK10; ser/Thr protein kinase PAR-1; Serine/threonine-protein kinase p78
Rabbit
Affinity chromatography
RUO
170577, 17169, 4140
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB3795
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P27448, Q03141, Q8VHF0
MARK3
A synthesized peptide derived from human MARK3 (aa 550-600).
100 μL
Primary
Human, Mouse, Rat