missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MARK3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12932-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is activated by phosphorylation and in turn is involved in the phosphorylation of tau proteins MAP2 and MAP4. Several transcript variants encoding different isoforms have been found for this gene.Specifications
| MARK3 | |
| Polyclonal | |
| Unconjugated | |
| MARK3 | |
| C TAK1, CTAK1, ELKL motif kinase 2, EMK 2, EMK2, KP78, MARK3, PAR1A, Protein kinase STK10, Ser/Thr protein kinase PAR 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 170577, 17169, 4140 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P27448, Q03141, Q8VHF0 | |
| Mark3 | |
| MARK3 Fusion Protein Ag3805 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction